Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003150.3 | NP_003141.2 | 24 | 4053 | ||
NM_139276.2 | NP_644805.1 | 24 | 4078 | |||
NM_213662.1 | NP_998827.1 | 24 | 3920 | |||
XM_011525145.2 | XP_011523447.1 | 24 | 4069 | |||
XM_011525146.2 | XP_011523448.1 | 24 | 4019 | |||
XM_017024973.1 | XP_016880462.1 | 24 | 4016 | |||
GenBank mRNA | BC008044.2 | - | 1 | 550 | ||
BC067119.1 | - | 1 | 198 | |||
BC107775.1 | - | 1 | 405 | |||
CF454565.1 | - | 1 | 196 |