Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEKT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053285.1 | NP_444515.1 | 2 | 246 | ||
XM_011524027.2 | XP_011522329.1 | 2 | 242 | |||
GenBank mRNA | AF357879.1 | - | 2 | 195 | ||
AK292818.1 | - | 2 | 211 | |||
AK304345.1 | - | 2 | 211 | |||
BC014599.2 | - | 2 | 246 | 4824860 |