Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052932.2 | NP_443164.2 | 4 | 1012 | ||
GenBank mRNA | AK075420.1 | - | 4 | 796 | ||
AK225790.1 | - | 4 | 983 | |||
AY008283.1 | - | 3 | 750 | |||
AY157580.1 | - | 4 | 707 | |||
AY358696.1 | - | 4 | 684 | |||
BC032296.1 | - | 4 | 699 | 4340763 |