Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSGA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304968.1 | NP_001291897.1 | 6 | 1093 | ||
NM_052933.3 | NP_443165.1 | 5 | 698 | |||
GenBank mRNA | AF294347.1 | - | 5 | 416 | ||
AF431815.1 | - | 5 | 698 | |||
AK093329.1 | - | 6 | 1092 | |||
BC038415.1 | - | 5 | 698 | 5172129 |