siRNA Details
Gene Symbol: TOP1MT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258446.1 | NP_001245375.1 | 4 | 372 | ||
NM_001258447.1 | NP_001245376.1 | 3 | 231 | |||
NM_052963.2 | NP_443195.1 | 3 | 280 | |||
XM_005250781.3 | XP_005250838.1 | 3 | 268 | |||
GenBank mRNA | AF349017.1 | - | 3 | 269 | ||
AK302372.1 | - | 3 | 231 | |||
AK303237.1 | - | 3 | 241 | |||
AK308475.1 | - | 3 | 252 | |||
AK309005.1 | - | 3 | 389 | |||
AK311647.1 | - | 3 | 252 | |||
AK315913.1 | - | 3 | 242 | |||
AK316380.1 | - | 4 | 319 | |||
BC044646.1 | - | 3 | 255 | |||
BC052285.1 | - | 3 | 255 | 5443410 | ||
BC071914.1 | - | 3 | 253 | 6372930 | ||
CB149603.1 | - | 3 | 270 | |||
DC375680.1 | - | 3 | 280 | |||
DC399840.1 | - | 3 | 231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
