Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TADA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053053.3 | NP_444281.1 | 2, 3 | 351 | ||
GenBank mRNA | AK290964.1 | - | 2, 3 | 168 | ||
AK291922.1 | - | 2, 3 | 291 | |||
AK309250.1 | - | 2, 3 | 219 | |||
BC015401.2 | - | 2, 3 | 176 | 4182025 | ||
CN399642.1 | - | 2, 3 | 353 |