Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSEN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127394.3 | NP_001120866.1 | 4 | 615 | ||
NM_001300764.1 | NP_001287693.1 | 5 | 769 | |||
NM_001300766.1 | NP_001287695.1 | 5 | 653 | |||
NM_052965.3 | NP_443197.1 | 5 | 757 | |||
NR_023349.2 | 6 | 874 | ||||
NR_125335.1 | 6 | 837 | ||||
GenBank mRNA | AF288394.1 | - | 5 | 732 | ||
AK098196.1 | - | 5 | 655 | |||
AK296655.1 | - | 4 | 510 | |||
AK307241.1 | - | 6 | 765 | |||
BC022030.1 | - | 5 | 702 | 4824025 | ||
BI821111.1 | - | 5 | 578 | |||
BM887891.1 | - | 4 | 447 |