Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006763.2 | NP_006754.1 | 2 | 2215 | ||
GenBank mRNA | U72649.1 | - | 2 | 2214 |