Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCGF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007144.2 | NP_009075.1 | 10 | 860 | ||
XM_005257640.2 | XP_005257697.1 | 11 | 1319 | |||
XM_005257641.4 | XP_005257698.1 | 9 | 1541 | |||
XM_005257642.3 | XP_005257699.1 | 9 | 1938 | |||
XM_017025016.1 | XP_016880505.1 | 11 | 1879 | |||
GenBank mRNA | BC004858.2 | - | 8 | 631 | 3841545 | |
BC024255.1 | - | 10 | 856 | 3605568 | ||
BX647429.1 | - | 11 | 1319 | |||
D13969.1 | - | 9 | 818 |