Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSANTD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198805.1 | NP_001185734.1 | 2 | 449 | ||
NM_001198806.1 | NP_001185735.1 | 2 | 456 | |||
NM_001198807.1 | NP_001185736.1 | 2 | 488 | |||
NM_080655.2 | NP_542386.1 | 2 | 488 | |||
GenBank mRNA | AK092292.1 | - | 1 | 64 | ||
AK314924.1 | - | 2 | 348 | |||
AY598327.1 | - | 1 | 274 | |||
BC008993.1 | - | 2 | 341 | 4213591 | ||
BI596498.1 | - | 2 | 446 | |||
BI600290.1 | - | 2 | 428 | |||
BM914752.1 | - | 2 | 367 | |||
CN366267.1 | - | 2 | 446 | |||
DA424604.1 | - | 2 | 456 | |||
DR159535.1 | - | 2 | 488 |
Gene Symbol: MSANTD3-TMEFF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198812.1 | NP_001185741.1 | 1 | 307 |