Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRSN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080723.4 | NP_542454.3 | 3 | 380 | ||
GenBank mRNA | AF418980.1 | - | 3 | 310 | ||
AK096290.1 | - | 3 | 457 | |||
AK225775.1 | - | 3 | 326 | |||
AK314373.1 | - | 3 | 348 | |||
AL833257.1 | - | 3 | 304 | |||
BC023514.2 | - | 3 | 307 | 4128341 |