Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080284.2 | NP_525023.2 | 10 | 1517 | ||
XM_011524574.2 | XP_011522876.1 | 4 | 845 | |||
XM_017024404.1 | XP_016879893.1 | 10 | 1829 | |||
XM_017024405.1 | XP_016879894.1 | 10 | 1829 | |||
XM_017024406.1 | XP_016879895.1 | 6 | 797 | |||
XM_017024407.1 | XP_016879896.1 | 7 | 901 | |||
GenBank mRNA | AF373290.1 | - | 9 | 1345 | ||
AK308431.1 | - | 10 | 1537 | |||
AY028898.1 | - | 10 | 1517 | |||
BC125231.1 | - | 10 | 1516 | |||
BC125232.1 | - | 10 | 1516 |