Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080650.3 | NP_542381.1 | 9 | 1249 | ||
GenBank mRNA | AK094007.1 | - | 9 | 1208 | ||
BC008485.2 | - | 8 | 1165 | |||
BC066652.1 | - | 9 | 1196 | 6066017 |