Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146225.1 | NP_001139697.1 | 2 | 699 | ||
NM_001146226.1 | NP_001139698.1 | 2 | 699 | |||
NM_080747.2 | NP_542785.1 | 2 | 699 | |||
XM_017018833.1 | XP_016874322.1 | 4 | 502 | |||
XM_017018834.1 | XP_016874323.1 | 4 | 440 | |||
XM_017018835.1 | XP_016874324.1 | 4 | 427 | |||
XM_017018836.1 | XP_016874325.1 | 3 | 401 | |||
GenBank mRNA | AK093060.1 | - | 2 | 603 | ||
AK293645.1 | - | 2 | 699 | |||
AK302016.1 | - | 3 | 734 | |||
AY033495.1 | - | 2 | 669 | |||
BC113686.1 | - | 2 | 633 | 8322738 |