Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322818.1 | NP_001309747.1 | 1, 2 | 152 | ||
XM_017013048.1 | XP_016868537.1 | 1, 2 | 1599 | |||
XM_017013049.1 | XP_016868538.1 | 1, 2 | 1550 | |||
XM_017013050.1 | XP_016868539.1 | 2, 3 | 1006 | |||
GenBank mRNA | AB055056.1 | - | 1, 2 | 151 |