Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130782.2 | NP_570138.1 | 1 | 269 | ||
GenBank mRNA | AF076642.1 | - | 1 | 243 | ||
AF268036.1 | - | 1 | 255 | |||
AK057114.1 | - | 1 | 262 | |||
AK292015.1 | - | 1 | 261 | |||
AK315377.1 | - | 1 | 262 | |||
AV715708.1 | - | 1 | 275 | |||
BC020632.1 | - | 1 | 261 | 4714909 |