Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080874.3 | NP_543150.1 | 1 | 280 | ||
XM_005262759.1 | XP_005262816.1 | 3 | 709 | |||
GenBank mRNA | AK096441.1 | - | 1 | 182 | ||
AY057053.1 | - | 1 | 166 | |||
BC065710.1 | - | 1 | 187 | 4272258 | ||
BX647857.1 | - | 1 | 281 |