Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMOT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113490.1 | NP_001106962.1 | 11 | 4978 | ||
NM_133265.2 | NP_573572.1 | 12 | 4547 | |||
XM_005262087.1 | XP_005262144.1 | 14 | 5723 | |||
XM_005262090.1 | XP_005262147.1 | 11 | 4220 | |||
XM_011530875.2 | XP_011529177.1 | 15 | 5835 | |||
XM_017029289.1 | XP_016884778.1 | 13 | 4751 | |||
GenBank mRNA | AB028994.3 | - | 10 | 3751 | ||
AF286598.1 | - | 12 | 4547 | |||
AK024344.1 | - | 1 | 688 | |||
BC050581.1 | - | 1 | 673 |