Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTX3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138287.3 | NP_612144.1 | 1 | 168 | ||
GenBank mRNA | AF484416.1 | - | 1 | 79 | ||
AK125086.1 | - | 1 | 168 | |||
AL833598.1 | - | 1 | 101 | |||
AY225123.1 | - | 1 | 213 | |||
AY780792.1 | - | 1 | 79 | |||
BC042191.1 | - | 1 | 87 | 4339554 | ||
BC060509.1 | - | 1 | 110 | 6457163 |