Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163424.1 | NP_001156896.1 | 2 | 165 | ||
NM_138336.1 | NP_612209.1 | 2 | 212 | |||
XM_017003411.1 | XP_016858900.1 | 2 | 637 | |||
GenBank mRNA | AF453951.1 | - | 2 | 212 | ||
AF487338.1 | - | 2 | 143 | |||
CN304277.1 | - | 2 | 149 |