Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270680.1 | NP_001257609.1 | 5 | 903 | ||
NM_138357.2 | NP_612366.1 | 5 | 626 | |||
NR_073062.1 | 7 | 791 | ||||
XM_017016882.1 | XP_016872371.1 | 5 | 826 | |||
GenBank mRNA | AK075439.1 | - | 5 | 614 | ||
AK128016.1 | - | 5 | 903 | |||
AK301475.1 | - | 7 | 772 | |||
AK310455.1 | - | 8 | 796 | |||
AK311133.1 | - | 7 | 785 | |||
AK315519.1 | - | 5 | 609 | |||
BC034235.1 | - | 5 | 626 | 5296557 |