Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MVB12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304547.1 | NP_001291476.1 | 9, 10 | 980 | ||
NM_138401.3 | NP_612410.1 | 8, 9 | 1053 | |||
GenBank mRNA | AK057057.1 | - | 9, 10 | 1538 | ||
AK289487.1 | - | 8, 9 | 853 | |||
BC007883.2 | - | 8, 9 | 789 | 3688605 | ||
BC011840.2 | - | 8, 9 | 797 | 4053084 | ||
BP394542.1 | - | 1, 2 | 27 | |||
BY996463.1 | - | 6, 7 | 614 |