Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136203.1 | NP_001129675.1 | 2 | 173 | ||
NM_138442.3 | NP_612451.1 | 2 | 235 | |||
GenBank mRNA | BC013949.2 | - | 2 | 235 | 2820921 | |
BG829184.1 | - | 2 | 239 | |||
BP309483.1 | - | 1 | 300 | |||
BQ669035.1 | - | 2 | 173 | |||
DB503140.1 | - | 2 | 173 |