Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MIPOL1 SIRNA Assays ›
Gene Symbol:
MIPOL1
Gene Name:
mirror-image polydactyly 1
Gene Aliases:
CCDC193
Chromosome Location:
Chr.14: 37197888 - 37579207 on Build GRCh38
UniGene ID:
Hs.660396
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
36773
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MIPOL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001195296.1
NP_001182225.1
6
836
NM_001195297.1
NP_001182226.1
6
822
NM_138731.6
NP_620059.1
5
683
XM_011536463.2
XP_011534765.1
8
1069
XM_011536467.2
XP_011534769.1
8
1066
XM_017020991.1
XP_016876480.1
8
1069
XM_017020992.1
XP_016876481.1
7
932
XM_017020993.1
XP_016876482.1
7
942
XM_017020994.1
XP_016876483.1
4
587
XM_017020995.1
XP_016876484.1
6
803
XM_017020996.1
XP_016876485.1
6
819
XM_017020997.1
XP_016876486.1
5
692
XM_017020998.1
XP_016876487.1
8
1068
XM_017020999.1
XP_016876488.1
9
1144
XM_017021000.1
XP_016876489.1
5
628
XM_017021001.1
XP_016876490.1
6
767
XM_017021002.1
XP_016876491.1
2
218
XM_017021003.1
XP_016876492.1
8
1067
XM_017021004.1
XP_016876493.1
4
595
XM_017021005.1
XP_016876494.1
5
690
XM_017021006.1
XP_016876495.1
8
1067
XM_017021007.1
XP_016876496.1
8
1067
XM_017021008.1
XP_016876497.1
5
626
XM_017021009.1
XP_016876498.1
6
739
XM_017021010.1
XP_016876499.1
2
218
XM_017021011.1
XP_016876500.1
4
551
XM_017021012.1
XP_016876501.1
6
739
XM_017021013.1
XP_016876502.1
2
218
XM_017021014.1
XP_016876503.1
8
1066
XM_017021015.1
XP_016876504.1
4
550
XM_017021016.1
XP_016876505.1
8
1066
XM_017021017.1
XP_016876506.1
8
1066
XM_017021018.1
XP_016876507.1
8
1066
GenBank mRNA
AK291850.1
-
6
747
AY059470.1
-
6
647
BC036124.1
-
6
708
5298587
BC036523.1
-
5
551
4828885
BX537396.1
-
5
646
BX537870.1
-
1
32
BX647795.1
-
5
522
CB105473.1
-
4
600
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0