Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM4SF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184723.1 | NP_001171652.1 | 1 | 252 | ||
NM_138786.3 | NP_620141.1 | 2 | 428 | |||
GenBank mRNA | AK313400.1 | - | 2 | 202 | ||
AL833134.1 | - | 2 | 351 | |||
BC014339.1 | - | 2 | 304 | 3828466 | ||
BF213446.1 | - | 263 | ||||
CA454331.1 | - | 1 | 214 |