Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136203.1 | NP_001129675.1 | 5 | 951 | ||
NM_138442.3 | NP_612451.1 | 5 | 1013 | |||
GenBank mRNA | AF086307.1 | - | 1 | 386 | ||
BC013949.2 | - | 5 | 1014 | 2820921 |