Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138703.4 | NP_619648.1 | 1 | 473 | ||
GenBank mRNA | AF490509.1 | - | 1 | 455 | ||
AK094624.1 | - | 1 | 465 | |||
BC093964.1 | - | 1 | 374 | 7939809 | ||
BC111949.1 | - | 1 | 374 | 8327417 | ||
DA793762.1 | - | 1 | 473 |