Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYTL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163334.1 | NP_001156806.1 | 2 | 541 | ||
NM_001163335.1 | NP_001156807.1 | 4 | 719 | |||
NM_138780.2 | NP_620135.1 | 3 | 628 | |||
XM_011544001.2 | XP_011542303.1 | 5 | 980 | |||
XM_011544002.2 | XP_011542304.1 | 3 | 614 | |||
XM_017029972.1 | XP_016885461.1 | 6 | 1146 | |||
XM_017029973.1 | XP_016885462.1 | 5 | 1213 | |||
XM_017029974.1 | XP_016885463.1 | 4 | 841 | |||
GenBank mRNA | AB080222.1 | - | 2 | 185 | ||
AK125106.1 | - | 3 | 597 | |||
BC131585.1 | - | 2 | 185 | 40113205 | ||
BX647688.1 | - | 4 | 721 |