Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HAUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138443.3 | NP_612452.1 | 5 | 658 | ||
NR_026978.1 | 5 | 698 | ||||
GenBank mRNA | AK024747.1 | - | 2 | 121 | ||
AK097403.1 | - | 5 | 639 | |||
AK315605.1 | - | 5 | 604 | |||
AY360137.1 | - | 5 | 658 | |||
BC005958.1 | - | 5 | 634 | 4047707 | ||
BC014003.2 | - | 5 | 631 | 3844806 |