Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276373.1 | NP_001263302.1 | 4, 5 | 293 | ||
NM_007122.4 | NP_009053.1 | 4, 5 | 389 | |||
GenBank mRNA | AK314876.1 | - | 4, 5 | 309 | ||
AL832119.1 | - | 4, 5 | 426 | |||
BC035505.2 | - | 4, 5 | 337 | 4559819 | ||
BP280296.1 | - | 4, 5 | 387 | |||
KU178379.1 | - | 2, 3 | 182 | |||
X55666.1 | - | 4, 5 | 302 |