Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUPT16H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007192.3 | NP_009123.1 | 2 | 417 | ||
XM_011536381.2 | XP_011534683.1 | 2 | 170 | |||
GenBank mRNA | AK001719.1 | - | 2 | 153 | ||
AK024072.1 | - | 2 | 419 | |||
BC000565.2 | - | 2 | 154 | |||
BC014046.2 | - | 2 | 154 | |||
BC064561.1 | - | 2 | 142 | |||
BC073849.1 | - | 2 | 151 | |||
BC111402.1 | - | 2 | 166 |