Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317192.1 | NP_001304121.1 | 3 | 260 | ||
NM_001317193.1 | NP_001304122.1 | 2 | 209 | |||
NM_001317194.1 | NP_001304123.1 | 3 | 260 | |||
NM_007241.3 | NP_009172.2 | 3 | 260 | |||
NR_133679.1 | 3 | 260 | ||||
XM_017024075.1 | XP_016879564.1 | 3 | 202 | |||
GenBank mRNA | AF156102.1 | - | 3 | 209 | ||
BC008976.1 | - | 3 | 210 | 3452153 | ||
BC038830.1 | - | 3 | 234 | 3900806 | ||
BE394098.1 | - | 3 | 163 | |||
BE818533.1 | - | 2 | 63 | |||
BG248541.1 | - | 3 | 249 | |||
BP342490.1 | - | 3 | 260 |