Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286633.1 | NP_001273562.1 | 4 | 1036 | ||
NM_138700.4 | NP_619645.1 | 3 | 867 | |||
XM_011514305.1 | XP_011512607.1 | 4 | 1467 | |||
XM_011514306.1 | XP_011512608.1 | 5 | 1333 | |||
XM_011514308.1 | XP_011512610.1 | 4 | 1467 | |||
XM_011514309.1 | XP_011512611.1 | 4 | 1467 | |||
GenBank mRNA | AF489517.1 | - | 3 | 590 | ||
BC060785.1 | - | 4 | 1036 | 30340682 |