Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139170.2 | NP_631909.2 | 2 | 554 | ||
XM_017022975.1 | XP_016878464.1 | 2 | 410 | |||
XM_017022976.1 | XP_016878465.1 | 2 | 172 | |||
GenBank mRNA | AF447587.1 | - | 2 | 528 | ||
AK301794.1 | - | 2 | 380 | |||
BC035024.1 | - | 2 | 504 | 4826923 | ||
BX648126.1 | - | 2 | 340 |