Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A6E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139249.2 | NP_640342.1 | 1 | 148 | ||
GenBank mRNA | AF333758.1 | - | 1 | 199 | ||
AF354931.1 | - | 1 | 148 | |||
AF354932.1 | - | 1 | 32 | |||
BC034588.1 | - | 1 | 62 | |||
BC069322.1 | - | 1 | 148 | 7262144 | ||
BC101783.1 | - | 1 | 208 | 8069289 |