Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAET1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243325.1 | NP_001230254.1 | 2 | 338 | ||
NM_001243327.1 | NP_001230256.1 | 2 | 446 | |||
NM_001243328.1 | NP_001230257.1 | 3 | 660 | |||
NM_139165.2 | NP_631904.1 | 2 | 446 | |||
XM_011535475.2 | XP_011533777.1 | 4 | 966 | |||
XM_011535476.2 | XP_011533778.1 | 4 | 980 | |||
XM_011535478.2 | XP_011533780.1 | 4 | 963 | |||
XM_011535479.2 | XP_011533781.1 | 4 | 965 | |||
XM_017010285.1 | XP_016865774.1 | 4 | 965 | |||
XM_017010286.1 | XP_016865775.1 | 4 | 965 | |||
XM_017010287.1 | XP_016865776.1 | 4 | 965 | |||
XM_017010288.1 | XP_016865777.1 | 2 | 214 | |||
GenBank mRNA | AF359243.1 | - | 2 | 334 | ||
AY054974.1 | - | 2 | 205 | |||
AY069961.1 | - | 2 | 373 | |||
AY176317.1 | - | 2 | 313 | |||
AY252119.1 | - | 2 | 313 | |||
AY359075.1 | - | 3 | 660 | |||
BC103694.1 | - | 2 | 344 | 40003751 | ||
EF489426.1 | - | 2 | 313 | |||
KJ628092.1 | - | 1 | 225 |