Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012425.3 | NP_036557.1 | 4 | 349 | ||
NM_152724.2 | NP_689937.2 | 3 | 237 | |||
XM_005252552.3 | XP_005252609.1 | 4 | 432 | |||
GenBank mRNA | AK225681.1 | - | 4 | 291 | ||
AK292911.1 | - | 3 | 162 | |||
AK312520.1 | - | 4 | 292 | |||
BC005993.1 | - | 4 | 319 | 4050837 | ||
BC008691.1 | - | 3 | 508 | 3872071 | ||
BC015644.2 | - | 4 | 309 | 4647836 | ||
BC107898.1 | - | 3 | 216 | |||
BG400752.1 | - | 3 | 238 | |||
BX647733.1 | - | 3 | 213 | |||
CR536521.1 | - | 3 | 206 | |||
CR541840.1 | - | 3 | 206 |