Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMA3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012431.2 | NP_036563.1 | 1 | 693 | ||
GenBank mRNA | AB002329.1 | - | 1 | 538 | ||
AK297209.1 | - | 1 | 373 | |||
BC140706.1 | - | 1 | 228 | 9021574 | ||
BC144338.1 | - | 1 | 228 | 9052858 | ||
BC152458.1 | - | 1 | 550 | 8862593 |