Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294340.1 | NP_001281269.1 | 2 | 388 | ||
NM_144604.3 | NP_653205.3 | 2 | 388 | |||
XM_011522863.1 | XP_011521165.1 | 2 | 388 | |||
XM_017022934.1 | XP_016878423.1 | 2 | 388 | |||
GenBank mRNA | AK225807.1 | - | 2 | 358 | ||
AK300254.1 | - | 2 | 366 | |||
AK302716.1 | - | 2 | 366 | |||
BC050463.1 | - | 2 | 335 | 6042036 | ||
CN299704.1 | - | 1 | 22 | |||
CN299707.1 | - | 2 | 347 | |||
DA313229.1 | - | 2 | 388 |