Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB5D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144607.4 | NP_653208.2 | 2 | 598 | ||
XM_005256440.3 | XP_005256497.1 | 2 | 377 | |||
XM_011523638.2 | XP_011521940.1 | 2 | 377 | |||
GenBank mRNA | AK057061.1 | - | 2 | 598 | ||
AK289520.1 | - | 2 | 281 | |||
BC060779.1 | - | 2 | 350 | 30349502 |