Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LOXHD1 SIRNA Assays ›
Gene Symbol:
LOXHD1
Gene Name:
lipoxygenase homology domains 1
Gene Aliases:
DFNB77, LH2D1
Chromosome Location:
Chr.18: 46476961 - 46657115 on Build GRCh38
UniGene ID:
Hs.345877
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
37281
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LOXHD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145472.2
NP_001138944.1
16
2454
NM_001145473.2
NP_001138945.1
2
315
NM_001173129.1
NP_001166600.1
2
315
NM_001308013.1
NP_001294942.1
14
2108
NM_144612.6
NP_653213.6
33
5192
XM_006722388.3
XP_006722451.1
15
2288
XM_006722389.3
XP_006722452.1
16
2464
XM_006722390.3
XP_006722453.1
16
2532
XM_006722391.3
XP_006722454.1
14
2102
XM_011525804.2
XP_011524106.1
22
3612
XM_011525807.2
XP_011524109.1
13
1998
XM_011525810.2
XP_011524112.1
2
535
XM_011525811.2
XP_011524113.1
2
1100
XM_017025540.1
XP_016881029.1
26
4408
XM_017025541.1
XP_016881030.1
27
4495
XM_017025542.1
XP_016881031.1
27
4495
XM_017025543.1
XP_016881032.1
27
4495
XM_017025544.1
XP_016881033.1
27
4495
XM_017025545.1
XP_016881034.1
27
4495
XM_017025548.1
XP_016881037.1
32
4842
GenBank mRNA
AK057232.1
-
2
302
AK127869.1
-
26
4505
AK302484.1
-
14
2099
BC041860.1
-
16
2436
5271046
BC047720.1
-
2
315
BX647229.1
-
14
2108
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
sensory perception of sound
detection of mechanical stimulus
calcium ion transmembrane transport
calcium channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0