Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178003.1 | NP_001171474.1 | 2 | 249 | ||
NM_144668.5 | NP_653269.3 | 2 | 236 | |||
GenBank mRNA | BC028421.2 | - | 2 | 219 | 4826893 | |
BC036233.1 | - | 2 | 214 | 5295406 | ||
BG717296.1 | - | 2 | 254 | |||
DB462471.1 | - | 2 | 232 |