Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RGS3 SIRNA Assays ›
Gene Symbol:
RGS3
Gene Name:
regulator of G-protein signaling 3
Gene Aliases:
C2PA, RGP3
Chromosome Location:
Chr.9: 113444729 - 113597743 on Build GRCh38
UniGene ID:
Hs.494875
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
37331
Validated
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RGS3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001276260.1
NP_001263189.1
4
1569
NM_001276261.1
NP_001263190.1
12
1493
NM_001276262.1
NP_001263191.1
2
217
NM_001282922.1
NP_001269851.1
5
1427
NM_001282923.1
NP_001269852.1
20
3037
NM_001322215.1
NP_001309144.1
4
1534
NM_130795.3
NP_570613.2
13
2471
NM_134427.2
NP_602299.1
2
491
NM_144488.5
NP_652759.3
23
3375
NM_144489.3
NP_652760.2
2
503
XM_006717219.2
XP_006717282.1
22
3221
XM_006717220.1
XP_006717283.1
20
2997
XM_006717222.1
XP_006717285.1
13
2347
XM_006717225.1
XP_006717288.1
4
1351
XM_006717226.2
XP_006717289.1
5
1644
XM_006717230.3
XP_006717293.1
4
1233
XM_011518894.1
XP_011517196.1
5
1536
XM_011518896.1
XP_011517198.1
5
1898
XM_011518897.2
XP_011517199.1
6
1896
XM_011518898.1
XP_011517200.1
6
1620
XM_011518900.1
XP_011517202.1
5
1421
XM_011518901.1
XP_011517203.1
5
1452
XM_017015003.1
XP_016870492.1
22
3250
XM_017015004.1
XP_016870493.1
19
2745
XM_017015005.1
XP_016870494.1
7
1584
XM_017015006.1
XP_016870495.1
5
1800
GenBank mRNA
AF463495.1
-
13
2471
AF490838.1
-
2
503
AF490839.1
-
20
2849
AF490840.1
-
13
2321
AF493927.1
-
3
1127
AF493941.1
-
3
188
AK054584.1
-
2
480
AK096840.1
-
12
1413
AK123387.1
-
4
1542
AK125059.1
-
20
3008
AK128127.1
-
23
3373
AK226153.1
-
3
2941
AK289666.1
-
5
1263
AK309971.1
-
1
1028
AY585192.1
-
3
1127
BC018072.1
-
2
294
BC019039.2
-
2
276
BC042555.1
-
13
2376
5175121
BE256545.1
-
2
217
BE298009.1
-
2
353
U27655.1
-
5
1414
Back To Top
More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Matching Taqman Gene Expression Assay:
Hs00367777_m1
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
inactivation of MAPK activity
regulation of G-protein coupled receptor protein signaling pathway
positive regulation of GTPase activity
GTPase activator activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0