Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144614.3 | NP_653215.2 | 2 | 236 | ||
GenBank mRNA | AF503919.2 | - | 2 | 237 |
Gene Symbol: LOC729458 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017027569.1 | XP_016883058.1 | 1 | 651 |
Gene Symbol: MBD3L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164425.1 | NP_001157897.1 | 2 | 231 |
Gene Symbol: MBD3L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164419.1 | NP_001157891.1 | 2 | 231 |
Gene Symbol: MBD3L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136507.1 | NP_001129979.1 | 2 | 231 |