Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RGS3 SIRNA Assays ›
Gene Symbol:
RGS3
Gene Name:
regulator of G-protein signaling 3
Gene Aliases:
C2PA, RGP3
Chromosome Location:
Chr.9: 113444729 - 113597743 on Build GRCh38
UniGene ID:
Hs.494875
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
37422
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RGS3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001276260.1
NP_001263189.1
7
2771
NM_001276261.1
NP_001263190.1
15
2695
NM_001276262.1
NP_001263191.1
5
1419
NM_001282922.1
NP_001269851.1
8
2629
NM_001282923.1
NP_001269852.1
23
4239
NM_001322215.1
NP_001309144.1
7
2736
NM_130795.3
NP_570613.2
16
3673
NM_134427.2
NP_602299.1
5
1693
NM_144488.5
NP_652759.3
26
4577
NM_144489.3
NP_652760.2
5
1705
XM_006717219.2
XP_006717282.1
25
4423
XM_006717220.1
XP_006717283.1
23
4199
XM_006717222.1
XP_006717285.1
16
3549
XM_006717225.1
XP_006717288.1
7
2553
XM_006717226.2
XP_006717289.1
8
2846
XM_006717230.3
XP_006717293.1
7
2435
XM_011518894.1
XP_011517196.1
8
2738
XM_011518896.1
XP_011517198.1
8
3100
XM_011518897.2
XP_011517199.1
9
3098
XM_011518898.1
XP_011517200.1
9
2822
XM_011518900.1
XP_011517202.1
8
2623
XM_011518901.1
XP_011517203.1
8
2654
XM_017015003.1
XP_016870492.1
25
4452
XM_017015004.1
XP_016870493.1
22
3947
XM_017015005.1
XP_016870494.1
10
2786
XM_017015006.1
XP_016870495.1
8
3002
GenBank mRNA
AF463495.1
-
16
3673
AK054584.1
-
5
1682
AK123387.1
-
7
2744
AK226153.1
-
6
4143
BC019039.2
-
5
1478
BC042555.1
-
16
3578
5175121
BM690084.1
-
1
239
U27655.1
-
8
2616
Back To Top
More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
inactivation of MAPK activity
regulation of G-protein coupled receptor protein signaling pathway
positive regulation of GTPase activity
GTPase activator activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0