Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077427.3 | NP_001070895.1 | 3 | 1000 | ||
NM_001321234.1 | NP_001308163.1 | 4 | 962 | |||
NM_001321235.1 | NP_001308164.1 | 3 | 1123 | |||
NM_144586.6 | NP_653187.3 | 3 | 1089 | |||
XM_011510546.2 | XP_011508848.1 | 4 | 1795 | |||
GenBank mRNA | AK075487.1 | - | 3 | 922 | ||
AK094501.1 | - | 4 | 865 | |||
AK130005.1 | - | 4 | 801 | |||
AK309668.1 | - | 3 | 1000 | |||
AK309849.1 | - | 4 | 809 | |||
AY358628.1 | - | 3 | 771 | |||
BC017318.2 | - | 3 | 902 | 3641660 |