Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOXHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145472.2 | NP_001138944.1 | 23 | 3470 | ||
NM_001145473.2 | NP_001138945.1 | 9 | 1331 | |||
NM_001173129.1 | NP_001166600.1 | 9 | 1331 | |||
NM_001308013.1 | NP_001294942.1 | 21 | 3124 | |||
NM_144612.6 | NP_653213.6 | 40 | 6208 | |||
XM_006722388.3 | XP_006722451.1 | 22 | 3304 | |||
XM_006722389.3 | XP_006722452.1 | 23 | 3480 | |||
XM_006722390.3 | XP_006722453.1 | 23 | 3548 | |||
XM_006722391.3 | XP_006722454.1 | 21 | 3118 | |||
XM_011525804.2 | XP_011524106.1 | 29 | 4628 | |||
XM_011525807.2 | XP_011524109.1 | 20 | 3014 | |||
XM_011525810.2 | XP_011524112.1 | 9 | 1551 | |||
XM_011525811.2 | XP_011524113.1 | 9 | 2116 | |||
XM_017025540.1 | XP_016881029.1 | 33 | 5424 | |||
XM_017025541.1 | XP_016881030.1 | 34 | 5511 | |||
XM_017025548.1 | XP_016881037.1 | 39 | 5858 | |||
GenBank mRNA | AK057232.1 | - | 9 | 1318 | ||
AK302484.1 | - | 21 | 3115 | |||
BC041860.1 | - | 23 | 3452 | 5271046 | ||
BC047720.1 | - | 9 | 1331 | |||
BX647229.1 | - | 21 | 3124 |