Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277074.1 | NP_001264003.1 | 3 | 238 | ||
NM_144716.5 | NP_653317.2 | 4 | 393 | |||
NR_102269.1 | 3 | 256 | ||||
XM_011533390.2 | XP_011531692.1 | 3 | 333 | |||
XM_011533391.2 | XP_011531693.1 | 4 | 399 | |||
XM_011533392.2 | XP_011531694.1 | 4 | 351 | |||
XM_011533393.2 | XP_011531695.1 | 2 | 1039 | |||
XM_011533394.2 | XP_011531696.1 | 3 | 875 | |||
XM_011533396.2 | XP_011531698.1 | 3 | 592 | |||
GenBank mRNA | AK096749.1 | - | 3 | 216 | ||
AK295532.1 | - | 3 | 216 | |||
AK303160.1 | - | 3 | 230 | |||
BC020830.1 | - | 3 | 219 | 4771328 | ||
BM803652.1 | - | 3 | 163 | |||
CN430023.1 | - | 4 | 348 | |||
CV574708.1 | - | 5 | 469 | |||
DA952452.1 | - | 256 | ||||
DC379144.1 | - | 3 | 241 |