Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF145 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199380.1 | NP_001186309.1 | 2 | 260 | ||
NM_001199381.1 | NP_001186310.1 | 2 | 307 | |||
NM_001199382.1 | NP_001186311.1 | 2 | 233 | |||
NM_001199383.1 | NP_001186312.1 | 2 | 384 | |||
NM_144726.2 | NP_653327.1 | 2 | 131 | |||
XM_017009138.1 | XP_016864627.1 | 2 | 579 | |||
GenBank mRNA | AK056513.1 | - | 2 | 131 | ||
AK292854.1 | - | 2 | 373 | |||
AK304228.1 | - | 2 | 219 | |||
AK304435.1 | - | 2 | 307 | |||
AK308394.1 | - | 2 | 260 | |||
AK311433.1 | - | 2 | 245 | |||
CK821118.1 | - | 2 | 399 | |||
DC418555.1 | - | 2 | 234 |