Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSTM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144984.3 | NP_659421.1 | 3 | 1141 | ||
GenBank mRNA | AK056299.1 | - | 3 | 1118 |